Learn More
Electron Microscopy Sciences Weigerts Iron Hematoxylin A 500 ML
Supplier: Electron Microscopy Sciences 2638602
Used in Goldner's Trichrome Method for connective tissue stains – Elastic tissue stain. Stain with a poly-acid step. See Luna, Lee G. Histopathologic Methods and Color Atlas of Special Stains and Tissue Artifacts. Johnson Printers, Downerts Grove, IL p151-152, c 1992.