Learn More
Electron Microscopy Sciences Weigerts Iron Hematoxylin Solution B 500 ML
SDP
Catalog No. 5031667
Encompass_Preferred
Used in Gridley's Method for Endamoeba Histolytica. For bacteria stain: Amoebae, their nuclei and ingested erythrocytes.
Catalog No. 50-316-67 Supplier Electron Microscopy Sciences Supplier No. 261091B
Used in Gridley's Method for Endamoeba Histolytica. For bacteria stain: Amoebae, their nuclei and ingested erythrocytes.