Learn More
Electron Microscopy Sciences Diamond Weigh Boats, 5 ml, Black, 35x 55 mm
SDP
Catalog No. 501929167
Encompass_Preferred
Our weigh boats are useful for liquid, powder or granular samples. They made of antistatic polystyrene and will withstand temperatures up to 90 deg, as well as exposure to diluted acids, alcohols, bases, and aqueous solutions. Their flexibility and smooth interior surfaces with rounded corners allow for easy decanting of powdered samples. Their flat bottoms ensure stability amd prevent tipping.
Originally designed as weighing containers, they have proven ideal for dispensing & storage and as disposable mixing/quick freezing trays.
Volume: 5 ml
Color: Black
Size: 35x 55 mm
Catalog No. 50-192-9167 Supplier Electron Microscopy Sciences Supplier No. 7001508
Our weigh boats are useful for liquid, powder or granular samples. They made of antistatic polystyrene and will withstand temperatures up to 90 deg, as well as exposure to diluted acids, alcohols, bases, and aqueous solutions. Their flexibility and smooth interior surfaces with rounded corners allow for easy decanting of powdered samples. Their flat bottoms ensure stability amd prevent tipping.
Originally designed as weighing containers, they have proven ideal for dispensing & storage and as disposable mixing/quick freezing trays.
Volume: 5 ml
Color: Black
Size: 35x 55 mm