Learn More
Sigma Aldrich Fine Chemicals Biosciences Weigh Boat Medium square b
Catalog No. 502901712
Encompass
flexible polystyrene boats feature smooth surfaces to provide accurate pour-outs with minimal sample loss.
Catalog No. 50-290-1712 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. HS120223
flexible polystyrene boats feature smooth surfaces to provide accurate pour-outs with minimal sample loss.