Learn More
Sigma Aldrich Fine Chemicals Biosciences Weigh Boat Medium square w
Catalog No. 502900470
Encompass
flexible polystyrene boats feature smooth surfaces to provide accurate pour-outs with minimal sample loss.
Catalog No. 50-290-0470 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. HS1420B
flexible polystyrene boats feature smooth surfaces to provide accurate pour-outs with minimal sample loss.