Learn More
Electron Microscopy Sciences Square Weigh Boats, 100 ml, Black, 80 x 80 mm
SDP
Catalog No. 501929163
Encompass_Preferred
Our weigh boats are useful for liquid, powder or granular samples. They made of antistatic polystyrene and will withstand temperatures up to 90 deg, as well as exposure to diluted acids, alcohols, bases, and aqueous solutions. Their flexibility and smooth interior surfaces with rounded corners allow for easy decanting of powdered samples. Their flat bottoms ensure stability amd prevent tipping.
Originally designed as weighing containers, they have proven ideal for dispensing & storage and as disposable mixing/quick freezing trays.
Volume: 100 ml
Color: Black
Size: 80 x 80 mm
Catalog No. 50-192-9163 Supplier Electron Microscopy Sciences Supplier No. 7001504
Our weigh boats are useful for liquid, powder or granular samples. They made of antistatic polystyrene and will withstand temperatures up to 90 deg, as well as exposure to diluted acids, alcohols, bases, and aqueous solutions. Their flexibility and smooth interior surfaces with rounded corners allow for easy decanting of powdered samples. Their flat bottoms ensure stability amd prevent tipping.
Originally designed as weighing containers, they have proven ideal for dispensing & storage and as disposable mixing/quick freezing trays.
Volume: 100 ml
Color: Black
Size: 80 x 80 mm