Learn More
Heathrow Scientific Weigh Boats
Catalog No. 01677188
Change view
Click to view available options
Size:
Small
Medium
Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
01-677-188 Large
01-677-186 Small
01-677-187 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-677-188 Supplier Heathrow Scientific Supplier No. 120126
Add to Cart
Add to Cart
Ensures accurate sample measurements of liquids, granules, and powders
  • Smooth, flat surfaces with easy-pour spouts for precise sample handling
  • Rounded corners for simplified transfer of samples
  • Constructed from durable, disposable polystyrene
  • Available in multiple colors and sizes
  • FDA/food-grade compliant
  • Biologically inert and resistant to dilute acids, alcohols, and bases
  • Suitable for use with microwave moisture analyzers
  • Shallow, wide, flat-bottom design for stability
  • Stackable for convenient storage
Specifications
Autoclavable Not Autoclavable
Bottom Shape Flat
Color White
Material Polystyrene
Shape Square
Sterility Non-sterile
Type Weigh Boat
Size Large
Width (English) 6 in.