Learn More
Andwin Scientific WEIGH BOATS LARGE 500/PK
SDP
Catalog No. NC9085517
Encompass_Preferred
The Heathrow Scientific Hexagonal Weighing Boats feature flat bottoms and sloping sides that give accurate transfer. The weighing boats are able to bend at points in order to form pour spouts. This product is in the large size and has an top diameter of 4" and base diameter of 3". It is sold in a pack of 500.
Catalog No. NC9085517 Supplier Andwin Scientific Supplier No. EF14251C
The Heathrow Scientific Hexagonal Weighing Boats feature flat bottoms and sloping sides that give accurate transfer. The weighing boats are able to bend at points in order to form pour spouts. This product is in the large size and has an top diameter of 4" and base diameter of 3". It is sold in a pack of 500.