Learn More
Cole-Parmer Cole-Parmer small Weigh Canoe with Pour Spout, Small, 2-1/4" L x 1-3/4" W x 5/16" D, 20 ml Capacity
Catalog No. NC9735287 Shop All Cole-Parmer Products
Encompass_Preferred
These polystyrene weigh dish facilitates pouring and dispensing. No need to flex dish to dispense liquids or solids; 20-30% thicker material for strength, stability, handling ease, and safety. New weigh dishes have a flat bottom and will withstand 200°F. They are disposable, smooth, uniform, and inexpensive. Available in three convenient sizes for any sample handling application.
Catalog No. NC9735287 Supplier Cole-Parmer Supplier No. 0101802
These polystyrene weigh dish facilitates pouring and dispensing. No need to flex dish to dispense liquids or solids; 20-30% thicker material for strength, stability, handling ease, and safety. New weigh dishes have a flat bottom and will withstand 200°F. They are disposable, smooth, uniform, and inexpensive. Available in three convenient sizes for any sample handling application.