Learn More
Andwin Scientific Weigh Dish 13 x 3.5
SDP
Catalog No. NC9310285
Encompass_Preferred
The aluminum micro weighing dish is suitable for microbalances, organic micro elementary and thermal analysis, and pharmaceutical use. This product has dimensions of 13dia. x 3.5D mm and has a capacity of 0.35 mL. These dishes can be used with most commercially available analyzers and microbalances. It is sold in a pack of 100.
Catalog No. NC9310285 Supplier Andwin Scientific Supplier No. 12577062
The aluminum micro weighing dish is suitable for microbalances, organic micro elementary and thermal analysis, and pharmaceutical use. This product has dimensions of 13dia. x 3.5D mm and has a capacity of 0.35 mL. These dishes can be used with most commercially available analyzers and microbalances. It is sold in a pack of 100.