Learn More
Cole-Parmer Cole-Parmer Essentials medium Square Polystyrene Weigh Boats, White, 100 mL, 100/Pk
Catalog No. NC2751787 Shop All Cole-Parmer Products
Encompass_Preferred
Rounded corners simplify transfer of powdered or granular samplesDisposableWhite Translucent Color Dishes - for traditional applicationsBlue Translucent Color Dishes - contrasts for white powders and solids, increases sample
Catalog No. NC2751787 Supplier Cole-Parmer Supplier No. 0101714
Rounded corners simplify transfer of powdered or granular samplesDisposableWhite Translucent Color Dishes - for traditional applicationsBlue Translucent Color Dishes - contrasts for white powders and solids, increases sample