Learn More
Cole-Parmer Cole-Parmer Essentials small Square Polystyrene Weigh Boats, White, 20 mL, 100/Pk
Catalog No. NC2751786 Shop All Cole-Parmer Products
Encompass_Preferred
No ReviewsWrite the First ReviewRounded corners simplify transfer of powdered or granular samplesDisposableWhite Translucent Color Dishes - for traditional applicationsBlue Translucent Color Dishes - contrasts for white powders and solids, increases sample
Catalog No. NC2751786 Supplier Cole-Parmer Supplier No. 0101704
No ReviewsWrite the First ReviewRounded corners simplify transfer of powdered or granular samplesDisposableWhite Translucent Color Dishes - for traditional applicationsBlue Translucent Color Dishes - contrasts for white powders and solids, increases sample