Learn More
Hach Company Weighing Boat 110x85mm
Supplier:  Hach Company 2179100
Encompass
Weighing Boat-85 Base DiameterFlexible plastic receptacles for handling liquids or solids during weighing These flexible polystyrene boats bend easily into a pouring spout to ensure non-spill transfer after weighing sample Stiff lip provides sure-grip surface for handling won t react to most weighing sample materials
Catalog No. 50-002-07452