Learn More
Hach Company Weighing Boat, 46 mm Base Diameter
Supplier:  Hach Company 2179000
Encompass
Flexible plastic receptacles for handling liquids or solids during weighing. These flexible polystyrene boats bend easily into a pouring spout to ensure non-spill transfer after weighing sample. Stiff lip provides sure-grip surface for handling; won’t react to most weighing sample materials.
Catalog No. 50-199-5508