Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Velp™ Weighing Boat for DK6 Digestion Unit
SDP

Catalog No. 11392189
Change view
Click to view available options
Dimensions (L x W x H):
70 x 23 x 15 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H)
11-392-189 70 x 23 x 15 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-392-189 Supplier Velp™ Supplier No. CM0486001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Weighing boats for use with the DK6 Digestion Unit.

  • Holds samples for digestion unit

Specifications

Autoclavable Not Autoclavable
Bottom Shape Flat
Color Clear
Disposable Single-use
Shape Rectangular
Sterility Non-sterile
Type Weighing Boat
For Use With (Equipment) DK6 Digestion Unit
No. per Pack 100/Pk.
Dimensions (L x W x H) 70 x 23 x 15 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.