Learn More
Research Products International Corp Disposable Weighing Dishes, Blue, 1-5/8 x 5/16 Inches, 500 per Package
SDP
Catalog No. 50212265
Encompass
For use with balances, microwave moisture analyzers and other sample handling applications. Rubberized polystyrene flexes for easy pouring. Biologically inert, resistant to dilute acids, aqueous solutions, alcohols, bases. Withstand temperatures up to 200°F. Flat bottom resists tipping. Contoured sides permit no-slip handling. Blue color contrasts with white powders and solids. Increases sample visibility and improves measuring accuracy.
Catalog No. 50-212-265 Supplier Research Products International Corp Supplier No. 143158
For use with balances, microwave moisture analyzers and other sample handling applications. Rubberized polystyrene flexes for easy pouring. Biologically inert, resistant to dilute acids, aqueous solutions, alcohols, bases. Withstand temperatures up to 200°F. Flat bottom resists tipping. Contoured sides permit no-slip handling. Blue color contrasts with white powders and solids. Increases sample visibility and improves measuring accuracy.