Learn More
Research Products International Corp Plastic Weighing Dishes, Natural, 5-1/2 x 7/8 Inches, 500 per Package
SDP
Catalog No. 50212262
Encompass
For use with balances, microwave moisture analyzers and other sample handling applications. Rubberized polystyrene flexes for easy pouring. Biologically inert, resistant to dilute acids, aqueous solutions, alcohols, bases. Withstand temperatures up to 200°F. Flat bottom resists tipping. Contoured sides permit no-slip handling.
Catalog No. 50-212-262 Supplier Research Products International Corp Supplier No. 142502
For use with balances, microwave moisture analyzers and other sample handling applications. Rubberized polystyrene flexes for easy pouring. Biologically inert, resistant to dilute acids, aqueous solutions, alcohols, bases. Withstand temperatures up to 200°F. Flat bottom resists tipping. Contoured sides permit no-slip handling.