Learn More
Sigma Aldrich Fine Chemicals Biosciences Weighing funnel, Small, 50/pk
Catalog No. NC1768403
Encompass
Sit flat on balance - No rolling, no spilling! Pouring necks designed to fit volumetric flasks, micro-tubes or any narrow neck apparatus for spill free transfer. Residual sample can be rinsed to guarantee 100% transfer. Suitable for safe handling of hazardous materials. Available in five different sizes: extra small, small, medium, large, and extra large.
Catalog No. NC1768403 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z5093371PAK
Sit flat on balance - No rolling, no spilling! Pouring necks designed to fit volumetric flasks, micro-tubes or any narrow neck apparatus for spill free transfer. Residual sample can be rinsed to guarantee 100% transfer. Suitable for safe handling of hazardous materials. Available in five different sizes: extra small, small, medium, large, and extra large.