Learn More
TWD Scientific LLC Small Polypropylene Weighing Funnel. Case of 1100
SDP
Catalog No. NC0235438 Shop All TWD Scientific LLC Products
Encompass_Preferred
Disposable polypropylene weighing funnel, size small. Fits volumetric flask volumes (mL) of 5, 10, 15, and 20. Funnels have a maximum capacity of 2mL Solid/2ml Liquid. 1100/Case
Catalog No. NC0235438 Supplier TWD Scientific LLC Supplier No. DPWFPP1S
Disposable polypropylene weighing funnel, size small. Fits volumetric flask volumes (mL) of 5, 10, 15, and 20. Funnels have a maximum capacity of 2mL Solid/2ml Liquid. 1100/Case