Learn More
Cole-Parmer Anti-static polypropylene weighing funnel, small
Catalog No. NC9884095 Shop All Cole-Parmer Products
Encompass_Preferred
Ensure complete sample transfer with these disposable polypropylene (PP) weighing funnels that are designed to fit in volumetric flasks. One side of the funnel is flattened so you can set the funnel flat on a balance. Micro funnel is designed to fit microwell plates. Autoclavable.
Catalog No. NC9884095 Supplier Cole-Parmer Supplier No. 0612702
Ensure complete sample transfer with these disposable polypropylene (PP) weighing funnels that are designed to fit in volumetric flasks. One side of the funnel is flattened so you can set the funnel flat on a balance. Micro funnel is designed to fit microwell plates. Autoclavable.