Learn More
Sigma Aldrich Fine Chemicals Biosciences Weighing funnel size XS an
Catalog No. 502901442
Encompass
Sit flat on balance - No rolling no spillingPouring necks designed to fit volumetric flasks micro-tubes or any narrow neck apparatus for spill free transferResidual sample can be rinsed to guarantee 100% transferSuitable for safe handling of hazardous materialsAvailable in five different sizes extra small small medium large and extra large
Catalog No. 50-290-1442 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z5093291PAK
Sit flat on balance - No rolling no spillingPouring necks designed to fit volumetric flasks micro-tubes or any narrow neck apparatus for spill free transferResidual sample can be rinsed to guarantee 100% transferSuitable for safe handling of hazardous materialsAvailable in five different sizes extra small small medium large and extra large