Learn More
Cole-Parmer Weighing Funnels, glass, large - fits 500 mL and 1 L flasks, 6/pk
Catalog No. NC0059792 Shop All Cole-Parmer Products
Encompass_Preferred
These reusable borosilicate glass weighing funnels are designed to ensure complete sample transfer to volumetric flasks. Funnels are flat on one side so you can set them on a balance. Micro funnel is sized to fit microwell plates.
Catalog No. NC0059792 Supplier Cole-Parmer Supplier No. 3470006
These reusable borosilicate glass weighing funnels are designed to ensure complete sample transfer to volumetric flasks. Funnels are flat on one side so you can set them on a balance. Micro funnel is sized to fit microwell plates.