Learn More
IW Tremont Co Inc LabExact Weighing paper, Nitrogen free, 4x4in unique analytical purity grade cellulose based paper for conveying powder or paste to balances and instruments, resists tearing during quadrant folding or moisture absorption. 500/pk
SDP
Catalog No. 502237585
Encompass_Preferred
LabExact This unique media is an analytical purity grade cellulose based paper designed for conveying powder or gelatinous paste samples to and from balances and instruments. The material has sufficient structural integrity to resist tearing during quadrant folding or enveloping. The surface is highly calendared (non treated gloss) to a smoothness which resists sample hold or moisture absorption. The surface texture is designed to increase abrasion durability during spatula scraping. This media demonstrates excellent lot-to-lot reproducibility. Material chemistries are verified against a standard using GC techniques prior to the paper making process. Other applications include; slip sheets between samples and slides, short term exposure as a moisture barrier, liners within containers and laminating backing for structural support.
This item is not returnable. View return policy
Catalog No. 50-223-7585 Supplier IW Tremont Co Inc Supplier No. LEW44
LabExact This unique media is an analytical purity grade cellulose based paper designed for conveying powder or gelatinous paste samples to and from balances and instruments. The material has sufficient structural integrity to resist tearing during quadrant folding or enveloping. The surface is highly calendared (non treated gloss) to a smoothness which resists sample hold or moisture absorption. The surface texture is designed to increase abrasion durability during spatula scraping. This media demonstrates excellent lot-to-lot reproducibility. Material chemistries are verified against a standard using GC techniques prior to the paper making process. Other applications include; slip sheets between samples and slides, short term exposure as a moisture barrier, liners within containers and laminating backing for structural support.