Learn More
Andwin Scientific WEIGHING PAPER 500/PK
SDP
Catalog No. NC0549964
Encompass_Preferred
The Weighing Paper is ideal for analytical weighing with minimal influence on analytical results for sample weighing and transporting. This product measures 7.6 x 7.6 cm (3" x 3"). It features a lightweight design, a smooth surface, and absence of du
Catalog No. NC0549964 Supplier Andwin Scientific Supplier No. 12578121
The Weighing Paper is ideal for analytical weighing with minimal influence on analytical results for sample weighing and transporting. This product measures 7.6 x 7.6 cm (3" x 3"). It features a lightweight design, a smooth surface, and absence of du