Learn More
PCE AMERICAS INC Laboratory Balance Scale with a resolution of 0.5 and a weighing capacity up to 30000 g 
SDP
Catalog No. NC3654891
Encompass_Preferred
PCE-WS 30 is an accurate and affordable digital tabletop scale with a resolution or readability of 0.5 and a weighing capacity up to 30000 g / 30.0 kg / 66.0 lb. Featuring adjustable feet and a built-in water bubble level, this benchtop electronic balance ensures precision with a full 100% tare weight range. PCE-WS 30 has a compact design and can be powered by the electric power adapter or the internal battery, making the scale portable when necessary.
Catalog No. NC3654891 Supplier PCE AMERICAS INC Supplier No. PCEWS 30
PCE-WS 30 is an accurate and affordable digital tabletop scale with a resolution or readability of 0.5 and a weighing capacity up to 30000 g / 30.0 kg / 66.0 lb. Featuring adjustable feet and a built-in water bubble level, this benchtop electronic balance ensures precision with a full 100% tare weight range. PCE-WS 30 has a compact design and can be powered by the electric power adapter or the internal battery, making the scale portable when necessary.