Learn More
Adam Equipment LBX 3 Latitude Compact Bench Scale, 6lb/3kg Capacity, 0.001lb / 0.5g Readability
Catalog No. NC0003644
Encompass_Preferred
Adam Equipments LBX compact bench scales deliver an excellent value for weighing, checkweighing, percentage weighing, and counting. With easy operation, Latitude is ideal for production, warehouse, or field use, as it is powered by the included AC adapter or internal rechargeable battery. The battery operates for 240 hours, so there are no worries about losing power. For added flexibility and convenience, Latitude can be charged using any USB power source, such as the USB connection in a vehicle or external USB power banks. With simple operation and a variety of modes, Latitude compact bench scales deliver an excellent value for quick, basic weighing and counting needs.Rechargeable battery included for operation almost anywhere.
Catalog No. NC0003644 Supplier Adam Equipment Supplier No. LBX 3
Adam Equipments LBX compact bench scales deliver an excellent value for weighing, checkweighing, percentage weighing, and counting. With easy operation, Latitude is ideal for production, warehouse, or field use, as it is powered by the included AC adapter or internal rechargeable battery. The battery operates for 240 hours, so there are no worries about losing power. For added flexibility and convenience, Latitude can be charged using any USB power source, such as the USB connection in a vehicle or external USB power banks. With simple operation and a variety of modes, Latitude compact bench scales deliver an excellent value for quick, basic weighing and counting needs.Rechargeable battery included for operation almost anywhere.