Learn More
Medicus Health Weighing Spatula, 150 Mm Long
SDP
Catalog No. NC1862870
Encompass
∙Weighing Spatulas transfer, lift, and handle powders and granular materials∙Great for breaking up clumps and releasing stuck powders in reagent bottles∙Made from nickel-plated, stainless steel wire∙Glazed spatula 5490M1 has a Hayman-type end∙Autoclavable∙Dimensions: Flat end, 30 mm L, Spoon, 12 mm L x 4 mm W - Glazed Finish
Catalog No. NC1862870 Supplier Medicus Health Supplier No. 5490M1
∙Weighing Spatulas transfer, lift, and handle powders and granular materials∙Great for breaking up clumps and releasing stuck powders in reagent bottles∙Made from nickel-plated, stainless steel wire∙Glazed spatula 5490M1 has a Hayman-type end∙Autoclavable∙Dimensions: Flat end, 30 mm L, Spoon, 12 mm L x 4 mm W - Glazed Finish