Learn More
Metrohm USA WEIGHING SPOON
SDP
Supplier:  Metrohm USA 62412000
Encompass
Weighing boat made of glass for the 6.1414.030 titration vessel lid or the 6.1455.31X, 6.1464.32X, 6.1465.320 titration vessel. For KF water determinations. Including protective tube.
Catalog No. NC0378118