Learn More
Rice Lake Weighing System Weight, 1lb ASTM Class 5
Catalog No. 50809365
Encompass_Preferred
Calibration Weight, Rice Lake Weighing Systems, Cylindrical, 1lb, ASTM Class 5, Stainless Steel, Satin finish, Density 7.84
Catalog No. 50-809-365 Supplier Rice Lake Weighing System Supplier No. 12596
Calibration Weight, Rice Lake Weighing Systems, Cylindrical, 1lb, ASTM Class 5, Stainless Steel, Satin finish, Density 7.84