Learn More
Rice Lake Weighing System Weight, 100g ASTM Class 5 
Catalog No. 50809344
Encompass_Preferred
Calibration Weight, Rice Lake Weighing Systems, Cylindrical, 100g, ASTM Class 5, Stainless Steel, Satin finish, Density 7.84
Catalog No. 50-809-344 Supplier Rice Lake Weighing System Supplier No. 12507
Calibration Weight, Rice Lake Weighing Systems, Cylindrical, 100g, ASTM Class 5, Stainless Steel, Satin finish, Density 7.84