Learn More
Rice Lake Weighing System Weight, 100mg ASTM Class 1
Catalog No. 50809322
Encompass_Preferred
Calibration Weight, Rice Lake Weighing Systems, Leaf, 100mg, ASTM, Class 1 SST Polished Density 7.95
Catalog No. 50-809-322 Supplier Rice Lake Weighing System Supplier No. 80461
Calibration Weight, Rice Lake Weighing Systems, Leaf, 100mg, ASTM, Class 1 SST Polished Density 7.95