Learn More
Rice Lake Weighing System WEIGHT 10MG LEAF ASTM CLASS1
Catalog No. 50809318
Encompass_Preferred
Calibration Weight, Rice Lake Weighing Systems, Leaf, 10mg, ASTM, Class 1 SST Polished Density 7.95
Catalog No. 50-809-318 Supplier Rice Lake Weighing System Supplier No. 11844
Calibration Weight, Rice Lake Weighing Systems, Leaf, 10mg, ASTM, Class 1 SST Polished Density 7.95