Learn More
Rice Lake Weighing System Weight, 200g ASTM Class 2
Catalog No. 50809415
Encompass_Preferred
Calibration Weight, Rice Lake Weighing Systems, Cylindrical, .2lb, ASTM Class 2, Stainless Steel, Satin finish, Density 7.84
Catalog No. 50-809-415 Supplier Rice Lake Weighing System Supplier No. 13067
Calibration Weight, Rice Lake Weighing Systems, Cylindrical, .2lb, ASTM Class 2, Stainless Steel, Satin finish, Density 7.84