Learn More
Rice Lake Weighing System Weight, 25lb ASTM Class 6 
Catalog No. 50809389
Encompass_Preferred
Calibration Weight, Rice Lake Weighing Systems, 25lb, Grip Handle, ASTM Class 6, Cast Iron, Painted
Catalog No. 50-809-389 Supplier Rice Lake Weighing System Supplier No. 12833
Calibration Weight, Rice Lake Weighing Systems, 25lb, Grip Handle, ASTM Class 6, Cast Iron, Painted