Learn More
Rice Lake Weighing System WEIGHT 30MG LEAF ASTM CLASS1
Catalog No. 50809320
Encompass_Preferred
Calibration Weight, Rice Lake Weighing Systems, Leaf, 30mg, ASTM, Class 1 SST Polished Density 7.95
Catalog No. 50-809-320 Supplier Rice Lake Weighing System Supplier No. 11846
Calibration Weight, Rice Lake Weighing Systems, Leaf, 30mg, ASTM, Class 1 SST Polished Density 7.95