Learn More
Rice Lake Weighing System Weight, 4oz ASTM Class 5
Catalog No. 50809362
Encompass_Preferred
Calibration Weight, Rice Lake Weighing Systems, Cylindrical, 4oz, ASTM Class 5, Stainless Steel, Satin finish, Density 7.84
Catalog No. 50-809-362 Supplier Rice Lake Weighing System Supplier No. 12592
Calibration Weight, Rice Lake Weighing Systems, Cylindrical, 4oz, ASTM Class 5, Stainless Steel, Satin finish, Density 7.84