Learn More
Rice Lake Weighing System Weight, 50g ASTM Class 5 
Catalog No. 50809354
Encompass_Preferred
Calibration Weight, Rice Lake Weighing Systems, Cylindrical, 50g ASTM Class 5, Stainless Steel, Satin finish, Density 7.84
Catalog No. 50-809-354 Supplier Rice Lake Weighing System Supplier No. 12527
Calibration Weight, Rice Lake Weighing Systems, Cylindrical, 50g ASTM Class 5, Stainless Steel, Satin finish, Density 7.84