Learn More
Rice Lake Weighing System Weight,Slotted Interlocking 50lb 10-1/2Dia x 2-1/2H x 9/16 Slot Width ASTM Class 6 Cast Iron Painted
Catalog No. 50809393
Encompass_Preferred
Calibration Weight, Rice Lake Weighing Systems, Slotted, Interlocking, 50lb, 10- 1/2 Dia x 2- 1/2 H x 9/16 Slot Width, ASTM Class 6, Cast Iron, Painted
Catalog No. 50-809-393 Supplier Rice Lake Weighing System Supplier No. 12856
Calibration Weight, Rice Lake Weighing Systems, Slotted, Interlocking, 50lb, 10- 1/2 Dia x 2- 1/2 H x 9/16 Slot Width, ASTM Class 6, Cast Iron, Painted