Learn More
Rice Lake Weighing System Weight, 5g ASTM Class 5 
Catalog No. 50809340
Encompass_Preferred
Calibration Weight, Rice Lake Weighing Systems, Cylindrical, 5g, ASTM Class 5, Stainless Steel, Satin finish, Density 7.84
Catalog No. 50-809-340 Supplier Rice Lake Weighing System Supplier No. 12503
Calibration Weight, Rice Lake Weighing Systems, Cylindrical, 5g, ASTM Class 5, Stainless Steel, Satin finish, Density 7.84