Learn More
Rice Lake Weighing System WEIGHT 5KG CAST INTRLOCK
Catalog No. 50809383
Encompass_Preferred
Calibration Weight, Rice Lake Weighing Systems, Slotted, Interlocking 5kg, 6- 5/32 Dia x 1- 25/32 H x 3/4 Slot Width, ASTM Class 7, Cast Iron, Painted
Catalog No. 50-809-383 Supplier Rice Lake Weighing System Supplier No. 12807
Calibration Weight, Rice Lake Weighing Systems, Slotted, Interlocking 5kg, 6- 5/32 Dia x 1- 25/32 H x 3/4 Slot Width, ASTM Class 7, Cast Iron, Painted