Learn More
Rice Lake Weighing System WEIGHT 5KG CYLINDRICAL CL F
Catalog No. 50809351
Encompass_Preferred
Calibration Weight, Rice Lake Weighing Systems, Cylindrical, 5kg NIST, Class F SST Satin Density 7.84
Catalog No. 50-809-351 Supplier Rice Lake Weighing System Supplier No. 12521
Calibration Weight, Rice Lake Weighing Systems, Cylindrical, 5kg NIST, Class F SST Satin Density 7.84