Learn More
Rice Lake Weighing System WEIGHT 5MG LEAF ASTM CLASS1
Catalog No. 50809317
Encompass_Preferred
Calibration Weight, Rice Lake Weighing Systems, Leaf, 5mg, ASTM, Class 1 SST Polished Density 7.95
Catalog No. 50-809-317 Supplier Rice Lake Weighing System Supplier No. 11842
Calibration Weight, Rice Lake Weighing Systems, Leaf, 5mg, ASTM, Class 1 SST Polished Density 7.95