Learn More
Andwin Scientific WEIGHT BOAT LARGE 500/PK
SDP
Catalog No. NC9387724
Encompass_Preferred
The Antistatic Polystyrene Weigh Boats are idea toolsl for weighing liquid or powdered samples in the laboratory. This product has a a smooth surface that gives accurate pour-outs with minimal sample loss and facilitates weighing of static-affected samples. The weigh boats are versatile, durable, economical, and secure. This product is sold in a pack of 500.
Catalog No. NC9387724 Supplier Andwin Scientific Supplier No. 12577049
The Antistatic Polystyrene Weigh Boats are idea toolsl for weighing liquid or powdered samples in the laboratory. This product has a a smooth surface that gives accurate pour-outs with minimal sample loss and facilitates weighing of static-affected samples. The weigh boats are versatile, durable, economical, and secure. This product is sold in a pack of 500.