Learn More
Cole-Parmer Cole-Parmer large Square Polystyrene Weigh Boats, Blue, 330 mL, 100/Pk
Catalog No. NC1125127 Shop All Cole-Parmer Products
Encompass_Preferred
General purpose sample holders. Translucent, rubberized polystyrene withstands temperatures to 180°F, flexes for easy pouring. Biologically inert; resistant to dilute acids, aqueous solutions, alcohols, bases. For use with balances, microwave moisture analyzers, hundreds of other sample handling applications. Flat bottom resists tipping. Contoured sides permit no-slip handling.
Catalog No. NC1125127 Supplier Cole-Parmer Supplier No. 0101726
General purpose sample holders. Translucent, rubberized polystyrene withstands temperatures to 180°F, flexes for easy pouring. Biologically inert; resistant to dilute acids, aqueous solutions, alcohols, bases. For use with balances, microwave moisture analyzers, hundreds of other sample handling applications. Flat bottom resists tipping. Contoured sides permit no-slip handling.