Learn More
Stellar Scientific Weigh Boats - Black, 5ml, 35 x55mm (Diamond) 500/CS
SDP
Catalog No. NC1143303
Encompass
Made from anti-static polystyrene and will withstand temperatures up to 90 C as well as exposure to diluted acids, alcohols, bases and aqueous solutions. Their flexibility and smooth interior surfaces with rounded corners allow for easy decanting of powdered samples. Their flat bottoms ensure stability and prevent tipping. Weigh boats are available in the traditional square shape as well as in a new diamond shape for easier pouring. Originally designed as weighing containers, they have proven ideal for dispensing and storage and as disposable mixing/quick freezing trays. Available in black for improved sample contrast/visibility and are packaged in ultra-compact dispenser packs.
Catalog No. NC1143303 Supplier Stellar Scientific Supplier No. WB05DIB
Made from anti-static polystyrene and will withstand temperatures up to 90 C as well as exposure to diluted acids, alcohols, bases and aqueous solutions. Their flexibility and smooth interior surfaces with rounded corners allow for easy decanting of powdered samples. Their flat bottoms ensure stability and prevent tipping. Weigh boats are available in the traditional square shape as well as in a new diamond shape for easier pouring. Originally designed as weighing containers, they have proven ideal for dispensing and storage and as disposable mixing/quick freezing trays. Available in black for improved sample contrast/visibility and are packaged in ultra-compact dispenser packs.