Learn More
Grainger RICE LAKE WEIGHING SYSTEMS, Calibration Weight, 10 lb Nominal
Catalog No. NC2610505
Encompass_Preferred
The Rice Lake Weighing Systems 10 lb Calibration Weight is a high-precision weight used for calibrating and checking scales for accuracy. It features a stainless steel construction with a knob handle on top for easy handling. The weight has an accuracy tolerance of +/- 5 grams to ensure reliable calibration. With NIST traceable calibration certificate, this 10 lb weight is ideal for use in industrial, laboratory, and other precision weighing applications. The durable build makes it suitable for daily use.
Catalog No. NC2610505 Supplier Grainger Supplier No. 15F061
The Rice Lake Weighing Systems 10 lb Calibration Weight is a high-precision weight used for calibrating and checking scales for accuracy. It features a stainless steel construction with a knob handle on top for easy handling. The weight has an accuracy tolerance of +/- 5 grams to ensure reliable calibration. With NIST traceable calibration certificate, this 10 lb weight is ideal for use in industrial, laboratory, and other precision weighing applications. The durable build makes it suitable for daily use.