Learn More
Rice Lake Weighing System WEIGHT KIT 100G ADJUSTED
Catalog No. 50809406
Encompass_Preferred
Calibration Weight Kit, Rice Lake Weighing Systems, 100g Adjusted, ASTM Class 1, SST Polished, Density 7.84, Electronic Balance Calibration Weights
Catalog No. 50-809-406 Supplier Rice Lake Weighing System Supplier No. 12946
Calibration Weight Kit, Rice Lake Weighing Systems, 100g Adjusted, ASTM Class 1, SST Polished, Density 7.84, Electronic Balance Calibration Weights