Learn More
Rice Lake Weighing System WEIGHT KIT 100G ASTM CLASS 2
Catalog No. 50809414
Encompass_Preferred
Calibration Weight Kit, Rice Lake Weighing Systems, 100g, ASTM Class 2, SST Satin, Density 7.84, Electronic Balance Calibration Weights
Catalog No. 50-809-414 Supplier Rice Lake Weighing System Supplier No. 13064
Calibration Weight Kit, Rice Lake Weighing Systems, 100g, ASTM Class 2, SST Satin, Density 7.84, Electronic Balance Calibration Weights