Learn More
Rice Lake Weighing System Weight Set,10lb-0.001lb ASTM Class 5
Catalog No. 50809375
Encompass_Preferred
Calibration Weight Kit, Rice Lake Weighing Systems,10lb-.001lb, ASTM Class 5, Stainless Steel, Satin finish, 31.61lb Kit
Catalog No. 50-809-375 Supplier Rice Lake Weighing System Supplier No. 12648
Calibration Weight Kit, Rice Lake Weighing Systems,10lb-.001lb, ASTM Class 5, Stainless Steel, Satin finish, 31.61lb Kit