Learn More
Rice Lake Weighing System WEIGHT KIT 1KG ADJUSTED ASTM
Catalog No. 50809412
Encompass_Preferred
Calibration Weight Kit, Rice Lake Weighing Systems, 1kg, Adjusted, ASTM Class 1, SST Polished, Density 7.84, Electronic Balance Calibration Weights
Catalog No. 50-809-412 Supplier Rice Lake Weighing System Supplier No. 12971
Calibration Weight Kit, Rice Lake Weighing Systems, 1kg, Adjusted, ASTM Class 1, SST Polished, Density 7.84, Electronic Balance Calibration Weights